siRNA Details
| Gene Symbol: Tmem40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168256.1 | NP_001161728.1 | 4 | 442 | ||
| NM_144805.2 | NP_659054.2 | 4 | 310 | |||
| GenBank mRNA | BC019416.1 | - | 4 | 272 | 5151159 | |
| CT010303.1 | - | 3 | 240 | |||
| CX564997.1 | - | 4 | 341 | |||

Tmem40
Genomic Map