siRNA Details
| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144815.2 | NP_659064.1 | 2 | 260 | ||
| GenBank mRNA | AK044228.1 | - | 2 | 460 | ||
| AK140387.1 | - | 2 | 281 | |||
| BC014705.1 | - | 2 | 269 | 4238201 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cecr5| Gene Symbol: Cecr5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144815.2 | NP_659064.1 | 2 | 260 | ||
| GenBank mRNA | AK044228.1 | - | 2 | 460 | ||
| AK140387.1 | - | 2 | 281 | |||
| BC014705.1 | - | 2 | 269 | 4238201 | ||