siRNA Details
| Gene Symbol: Ccdc92 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144819.2 | NP_659068.1 | 3, 4 | 404 | ||
| GenBank mRNA | AK028192.1 | - | 3, 4 | 381 | ||
| AK136203.1 | - | 3, 4 | 570 | |||
| AK165371.1 | - | 3, 4 | 404 | |||
| BC021492.1 | - | 3, 4 | 348 | 5345674 | ||

Ccdc92
Genomic Map