siRNA Details
| Gene Symbol: Ccdc28a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144820.3 | NP_659069.2 | 4 | 549 | ||
| GenBank mRNA | AK028075.1 | - | 4 | 518 | ||
| AK046998.1 | - | 2 | 173 | |||
| AK148447.1 | - | 4 | 550 | |||
| BC013553.1 | - | 4 | 547 | |||
| BC027759.1 | - | 4 | 557 | 5346317 | ||

Ccdc28a
Genomic Map