siRNA Details
| Gene Symbol: Heatr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144835.4 | NP_659084.3 | 39 | 5704 | ||
| GenBank mRNA | AK032134.1 | - | 4 | 629 | ||
| AK132192.1 | - | 1378 | ||||
| BC019693.1 | - | 3 | 343 | |||
| BC066225.1 | - | 1 | 795 | |||
| BC069833.1 | - | 1 | 795 | |||

Heatr1
Genomic Map