siRNA Details
| Gene Symbol: Slc17a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144836.2 | NP_659085.1 | 3 | 632 | ||
| GenBank mRNA | AK040717.1 | - | 3 | 633 | ||
| AK050184.1 | - | 3 | 615 | |||
| AK050277.1 | - | 4 | 596 | |||
| BC018306.1 | - | 2 | 270 | 4193755 | ||

Slc17a2
Genomic Map