siRNA Details
| Gene Symbol: Fam49b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144846.5 | NP_659095.1 | 4 | 425 | ||
| GenBank mRNA | AK028062.1 | - | 4 | 426 | ||
| AK075613.1 | - | 5 | 370 | |||
| AK183968.1 | - | 2 | 119 | |||
| AK205521.1 | - | 3 | 174 | |||
| BC011343.1 | - | 4 | 243 | 4012217 | ||

Fam49b
Genomic Map