siRNA Details
| Gene Symbol: Cyyr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144853.3 | NP_659102.1 | 3 | 508 | ||
| GenBank mRNA | AF442733.1 | - | 3 | 243 | ||
| AK031840.1 | - | 3 | 532 | |||
| AY061854.1 | - | 3 | 530 | |||
| BC099957.1 | - | 3 | 508 | 6431603 | ||
| BC125594.1 | - | 3 | 302 | 40130109 | ||
| BC125596.1 | - | 3 | 302 | 40130111 | ||

Cyyr1
Genomic Map