siRNA Details
| Gene Symbol: Reep2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001204914.1 | NP_001191843.1 | 4, 5 | 472 | ||
| NM_144865.2 | NP_659114.2 | 4, 5 | 472 | |||
| GenBank mRNA | AY562230.1 | - | 4, 5 | 305 | ||
| BC020184.1 | - | 4, 5 | 403 | 4505679 | ||
| CX216997.1 | - | 4, 5 | 472 | |||
| EU555417.1 | - | 4, 5 | 305 | |||

Reep2
Genomic Map