siRNA Details
| Gene Symbol: Uhrf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144873.2 | NP_659122.2 | 2 | 541 | ||
| GenBank mRNA | AB116653.1 | - | 2 | 276 | ||
| AF274047.1 | - | 2 | 451 | |||
| AK031036.1 | - | 2 | 603 | |||
| AK041564.1 | - | 2 | 563 | |||
| AK042321.1 | - | 2 | 582 | |||
| AK051743.1 | - | 2 | 582 | |||
| AK080925.1 | - | 2 | 582 | |||
| AK172682.1 | - | 2 | 425 | |||
| AK184966.1 | - | 1 | 90 | |||
| BC060241.1 | - | 2 | 541 | 6334856 | ||

Uhrf2
Genomic Map