siRNA Details
| Gene Symbol: Proser2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159657.1 | NP_001153129.1 | 4 | 1204 | ||
| NM_144883.4 | NP_659132.2 | 4 | 1058 | |||
| GenBank mRNA | AK077380.1 | - | 4 | 1034 | ||
| AK132370.1 | - | 1008 | ||||
| BC023193.1 | - | 3 | 917 | 4946741 | ||
| BC025867.1 | - | 5 | 1165 | 5052194 | ||
| BC026904.1 | - | 1 | 128 | |||
| BC029968.1 | - | 4 | 1021 | |||

Proser2
Genomic Map