siRNA Details
| Gene Symbol: Chpt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146690.1 | NP_001140162.1 | 2 | 480 | ||
| NM_144807.3 | NP_659056.3 | 2 | 480 | |||
| GenBank mRNA | AK007964.1 | - | 2 | 400 | ||
| AK032497.1 | - | 2 | 375 | |||
| AY445814.1 | - | 2 | 375 | |||
| BC016251.1 | - | 2 | 428 | 4911104 | ||
| BC031435.1 | - | 2 | 435 | 4221062 | ||
| EL958096.1 | - | 1 | 80 | |||

Chpt1
Genomic Map