siRNA Details
| Gene Symbol: Zmym5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001253752.1 | NP_001240681.1 | 8 | 1429 | ||
| NM_001253753.1 | NP_001240682.1 | 8 | 1499 | |||
| NM_144842.3 | NP_659091.3 | 8 | 1493 | |||
| GenBank mRNA | AK036513.1 | - | 7 | 1338 | ||
| AK155333.1 | - | 8 | 1411 | |||
| AK156501.1 | - | 8 | 1501 | |||
| AK165292.1 | - | 1 | 154 | |||
| BC027380.1 | - | 6 | 842 | |||
| CN534727.1 | - | 2 | 210 | |||

Zmym5
Genomic Map