siRNA Details
| Gene Symbol: Vash2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144879.2 | NP_659128.2 | 3 | 768 | ||
| GenBank mRNA | AK045261.1 | - | 3 | 564 | ||
| AK078596.1 | - | 3 | 637 | |||
| AK140989.1 | - | 3 | 762 | |||
| AK163193.1 | - | 3 | 544 | |||
| BC024141.1 | - | 3 | 538 | 5149750 | ||

Vash2
Genomic Map