siRNA Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144880.4 | NP_659129.2 | 13 | 2196 | ||
| GenBank mRNA | AK145397.1 | - | 14 | 1582 | ||
| BC023062.1 | - | 7 | 765 | |||
| BC034058.1 | - | 3 | 612 | |||
| BC059026.1 | - | 13 | 2195 | 6810754 | ||

Ppp2r5a
Genomic Map