siRNA Details
| Gene Symbol: Bpifb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144890.2 | NP_659139.2 | 2 | 276 | ||
| GenBank mRNA | AK142982.1 | - | 2 | 277 | ||
| BC018465.1 | - | 2 | 238 | 3156626 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bpifb5| Gene Symbol: Bpifb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144890.2 | NP_659139.2 | 2 | 276 | ||
| GenBank mRNA | AK142982.1 | - | 2 | 277 | ||
| BC018465.1 | - | 2 | 238 | 3156626 | ||