siRNA Details
| Gene Symbol: Spg20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144987.1 | NP_001138459.1 | 2 | 453 | ||
| NM_001144988.1 | NP_001138460.1 | 2 | 237 | |||
| NM_144895.2 | NP_659144.1 | 2 | 237 | |||
| GenBank mRNA | AK035081.1 | - | 1 | 96 | ||
| AK087388.1 | - | 2 | 204 | |||
| AK089946.1 | - | 2 | 237 | |||
| AK129171.1 | - | 2 | 216 | |||
| AK147760.1 | - | 2 | 454 | |||
| AK159951.1 | - | 2 | 238 | |||
| BC022921.1 | - | 2 | 205 | 4220506 | ||

Spg20
Genomic Map