siRNA Details
| Gene Symbol: Depdc7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144804.1 | NP_659053.1 | 8 | 1373 | ||
| GenBank mRNA | AK134830.1 | - | 8 | 1393 | ||
| AK144066.1 | - | 8 | 1386 | |||
| AK162086.1 | - | 8 | 1397 | |||
| AK162218.1 | - | 8 | 1396 | |||
| BC013499.1 | - | 8 | 1373 | 4222591 | ||

Depdc7
Genomic Map