siRNA Details
| Gene Symbol: Tmem40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168256.1 | NP_001161728.1 | 8, 9 | 654 | ||
| NM_001168257.1 | NP_001161729.1 | 7, 8 | 579 | |||
| NM_001168258.1 | NP_001161730.1 | 3, 4 | 379 | |||
| NM_001168259.1 | NP_001161731.1 | 4, 5 | 146 | |||
| NM_144805.2 | NP_659054.2 | 8, 9 | 522 | |||
| GenBank mRNA | AK078864.1 | - | 4, 5 | 147 | ||
| BC019416.1 | - | 8, 9 | 484 | 5151159 | ||
| BX511925.1 | - | 3, 4 | 106 | |||
| CT010303.1 | - | 7, 8 | 452 | |||
| CV561368.1 | - | 7, 8 | 521 | |||
| CX564997.1 | - | 8, 9 | 553 | |||

Tmem40
Genomic Map