siRNA Details
| Gene Symbol: Zfp410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144833.3 | NP_659082.1 | 12 | 1882 | ||
| GenBank mRNA | AF295806.1 | - | 12 | 1644 | ||
| AK051939.1 | - | 10 | 1282 | |||
| AK083003.1 | - | 12 | 1625 | |||
| AK132345.1 | - | 12 | 1621 | |||
| BC021528.1 | - | 12 | 1637 | 5353673 | ||
| BC056957.1 | - | 12 | 1579 | |||

Zfp410
Genomic Map