siRNA Details
| Gene Symbol: Otx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001286481.1 | NP_001273410.1 | 5 | 1381 | ||
| NM_001286482.1 | NP_001273411.1 | 5 | 1357 | |||
| NM_001286483.1 | NP_001273412.1 | 4 | 1197 | |||
| NM_144841.4 | NP_659090.1 | 3 | 1279 | |||
| GenBank mRNA | AK053741.1 | - | 4 | 1194 | ||
| AK081535.1 | - | 4 | 1198 | |||
| AK087527.1 | - | 4 | 1195 | |||
| BC017609.1 | - | 3 | 1279 | 4527414 | ||
| BC027104.1 | - | 5 | 1239 | 5359966 | ||
| BC029667.1 | - | 5 | 1243 | 5362013 | ||

Otx2
Genomic Map