siRNA Details
| Gene Symbol: Nrbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144847.1 | NP_659096.1 | 12 | 1847 | ||
| GenBank mRNA | BC011468.1 | - | 12 | 1780 | 4012880 | |
| BC012437.1 | - | 12 | 1847 | 3710089 | ||
| BC025042.1 | - | 7 | 1309 | |||
| BC068117.1 | - | 17 | 2495 | |||

Nrbp2
Genomic Map