siRNA Details
| Gene Symbol: Ankrd54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144849.1 | NP_659098.1 | 8 | 1411 | ||
| GenBank mRNA | AK050440.1 | - | 7 | 2126 | ||
| AK133785.1 | - | 8 | 1435 | |||
| AK143625.1 | - | 8 | 1439 | |||
| BC014743.1 | - | 8 | 1411 | 4503319 | ||
| DQ884878.1 | - | 8 | 1439 | |||

Ankrd54
Genomic Map