siRNA Details
| Gene Symbol: Wdr36 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110015.1 | NP_001103485.1 | 13 | 1603 | ||
| NM_001110016.1 | NP_001103486.1 | 13 | 1603 | |||
| NM_144863.4 | NP_659112.2 | 13 | 1603 | |||
| GenBank mRNA | AK041763.1 | - | 13 | 1523 | ||
| AK045396.1 | - | 13 | 1576 | |||
| AK077545.1 | - | 13 | 1532 | |||
| AK155965.1 | - | 13 | 1501 | |||
| AK156822.1 | - | 1602 | ||||
| AK171683.1 | - | 13 | 1541 | |||
| AK172054.1 | - | 13 | 1528 | |||
| AK172377.1 | - | 13 | 1533 | |||
| AK186642.1 | - | 2 | 198 | |||
| AK214226.1 | - | 2 | 198 | |||
| BC022132.1 | - | 13 | 1575 | |||
| BC025809.1 | - | 13 | 1517 | |||
| BC094453.1 | - | 13 | 1554 | 5068941 | ||

Wdr36
Genomic Map