siRNA Details
| Gene Symbol: Mettl13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144877.1 | NP_659126.1 | 8 | 2416 | ||
| GenBank mRNA | AK084750.1 | - | 8 | 2111 | ||
| AK146751.1 | - | 2614 | ||||
| AK168959.1 | - | 8 | 2623 | |||
| AK169319.1 | - | 2615 | ||||
| AK173044.1 | - | 2569 | ||||
| BC014872.1 | - | 8 | 2416 | 3602469 | ||

Mettl13
Genomic Map