siRNA Details
| Gene Symbol: Exosc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144886.2 | NP_659135.1 | 9 | 818 | ||
| GenBank mRNA | AK144234.1 | - | 9 | 827 | ||
| AK164618.1 | - | 8 | 1600 | |||
| AK166881.1 | - | 9 | 828 | |||
| BC021485.1 | - | 9 | 842 | 5344620 | ||
| BC021807.1 | - | 9 | 840 | 3980155 | ||

Exosc2
Genomic Map