siRNA Details
| Gene Symbol: Slc35a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144902.3 | NP_659151.1 | 3 | 498 | ||
| GenBank mRNA | AK029398.1 | - | 3 | 870 | ||
| AK041195.1 | - | 4 | 618 | |||
| AK049484.1 | - | 3 | 498 | |||
| AK161317.1 | - | 3 | 456 | |||
| BC024110.1 | - | 3 | 494 | 5150304 | ||

Slc35a3
Genomic Map