siRNA Details
| Gene Symbol: Rtbdn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144929.2 | NP_659178.1 | 3 | 1002 | ||
| GenBank mRNA | AK039726.1 | - | 1 | 1509 | ||
| AK080715.1 | - | 3 | 318 | |||
| AY428631.1 | - | 2 | 214 | |||
| AY527275.1 | - | 3 | 1002 | |||
| AY527276.1 | - | 4 | 457 | |||
| BC022744.1 | - | 2 | 574 | 4527012 | ||
| BC023051.1 | - | 2 | 530 | 5361063 | ||
| BC027079.1 | - | 2 | 530 | 5357069 | ||

Rtbdn
Genomic Map