siRNA Details
| Gene Symbol: Lppr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290299.1 | NP_001277228.1 | 4 | 482 | ||
| NM_144935.1 | NP_659184.1 | 3 | 454 | |||
| GenBank mRNA | AK082453.1 | - | 4 | 445 | ||
| AY438023.1 | - | 3 | 454 | |||
| BC018242.1 | - | 3 | 454 | 4503683 | ||
| BC023082.1 | - | 4 | 459 | 5357160 | ||
| CX122376.1 | - | 4 | 482 | |||

Lppr2
Genomic Map