siRNA Details
| Gene Symbol: 2610305D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011986.1 | - | 2 | 393 | ||
| BC032186.1 | - | 1 | 179 | |||
| BC138179.1 | - | 2 | 334 | 8861198 | ||
| BC145854.1 | - | 2 | 334 | 40131157 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2610305D13Rik| Gene Symbol: 2610305D13Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK011986.1 | - | 2 | 393 | ||
| BC032186.1 | - | 1 | 179 | |||
| BC138179.1 | - | 2 | 334 | 8861198 | ||
| BC145854.1 | - | 2 | 334 | 40131157 | ||