siRNA Details
| Gene Symbol: CD3E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000733.3 | NP_000724.1 | 6 | 474 | ||
| GenBank mRNA | AK292612.1 | - | 6 | 323 | ||
| AK301255.1 | - | 6 | 324 | |||
| AK311641.1 | - | 5 | 771 | |||
| BC049847.1 | - | 5 | 286 | 5214563 | ||
| BI910359.1 | - | 6 | 509 | |||
| BP366535.1 | - | 6 | 491 | |||
| X03884.1 | - | 5 | 272 | |||

CD3E
Genomic Map