siRNA Details
| Gene Symbol: Cd207 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144943.3 | NP_659192.2 | 2, 3 | 255 | ||
| GenBank mRNA | AJ302711.1 | - | 2, 3 | 255 | ||
| AK142731.1 | - | 2, 3 | 197 | |||
| AY026050.1 | - | 2, 3 | 197 | |||
| AY078415.1 | - | 2, 3 | 197 | |||

Cd207
Genomic Map