siRNA Details
| Gene Symbol: Trpv3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145099.2 | NP_659567.2 | 5 | 399 | ||
| GenBank mRNA | AB083654.1 | - | 5 | 375 | ||
| AF510316.1 | - | 5 | 399 | |||
| BC108984.1 | - | 5 | 385 | 40047664 | ||
| BC108985.1 | - | 5 | 385 | |||

Trpv3
Genomic Map