siRNA Details
| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1418 | ||
| GenBank mRNA | BI990274.1 | - | 1 | 357 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crebzf| Gene Symbol: Crebzf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145151.3 | NP_660133.2 | 1 | 1418 | ||
| GenBank mRNA | BI990274.1 | - | 1 | 357 | ||