siRNA Details
| Gene Symbol: 1700019D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144953.2 | NP_659202.1 | 2 | 807 | ||
| GenBank mRNA | AK010796.1 | - | 2 | 808 | ||
| AK010944.1 | - | 2 | 669 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700019D03Rik| Gene Symbol: 1700019D03Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_144953.2 | NP_659202.1 | 2 | 807 | ||
| GenBank mRNA | AK010796.1 | - | 2 | 808 | ||
| AK010944.1 | - | 2 | 669 | |||