siRNA Details
| Gene Symbol: Aif1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145144.1 | NP_660126.1 | 6 | 1441 | ||
| GenBank mRNA | AK028955.1 | - | 6 | 1399 | ||
| AK045539.1 | - | 6 | 1471 | |||
| BC024599.1 | - | 6 | 1441 | 5044495 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Aif1l| Gene Symbol: Aif1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145144.1 | NP_660126.1 | 6 | 1441 | ||
| GenBank mRNA | AK028955.1 | - | 6 | 1399 | ||
| AK045539.1 | - | 6 | 1471 | |||
| BC024599.1 | - | 6 | 1441 | 5044495 | ||