siRNA Details
| Gene Symbol: Oasl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145209.3 | NP_660210.1 | 7 | 1543 | ||
| GenBank mRNA | AB067533.1 | - | 6 | 1468 | ||
| AK078690.1 | - | 7 | 1544 | |||
| AK089111.1 | - | 6 | 1515 | |||
| AK149824.1 | - | 6 | 1467 | |||
| AK150863.1 | - | 6 | 1597 | |||
| AK165578.1 | - | 6 | 1580 | |||
| AK172183.1 | - | 2 | 1967 | |||
| AY057107.1 | - | 6 | 1456 | |||
| AY089728.1 | - | 6 | 1468 | |||

Oasl1
Genomic Map