siRNA Details
| Gene Symbol: Gps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177874.1 | NP_001171345.1 | 3 | 258 | ||
| NM_145370.2 | NP_663345.2 | 3 | 398 | |||
| GenBank mRNA | AK088490.1 | - | 3 | 255 | ||
| BC003350.1 | - | 3 | 398 | 3481979 | ||
| BC103782.1 | - | 3 | 225 | 6486599 | ||

Gps1
Genomic Map