siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145382.4 | NP_663357.2 | 7, 8 | 2772 | ||
| GenBank mRNA | AK040568.1 | - | 1, 2 | 900 | ||
| AK155237.1 | - | 7, 8 | 2766 | |||
| AK173303.1 | - | 7, 8 | 1621 | |||
| BC021381.1 | - | 1, 2 | 362 | 5043282 | ||
| BC023015.1 | - | 1, 2 | 304 | 5364758 | ||
| BC025483.1 | - | 7, 8 | 1631 | 5150988 | ||
| BC051255.1 | - | 1, 2 | 3220 | |||

Fam193b
Genomic Map