siRNA Details
| Gene Symbol: BC016579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145389.2 | NP_663364.2 | 3 | 439 | ||
| GenBank mRNA | AK078878.1 | - | 3 | 390 | ||
| BC016579.1 | - | 3 | 439 | 2615865 | ||
| CD552400.1 | - | 1 | 78 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

BC016579| Gene Symbol: BC016579 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145389.2 | NP_663364.2 | 3 | 439 | ||
| GenBank mRNA | AK078878.1 | - | 3 | 390 | ||
| BC016579.1 | - | 3 | 439 | 2615865 | ||
| CD552400.1 | - | 1 | 78 | |||