siRNA Details
| Gene Symbol: Slc25a29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_181328.3 | NP_851845.1 | 4 | 1282 | ||
| GenBank mRNA | AK047628.1 | - | 4 | 1283 | ||
| AK087057.1 | - | 4 | 1270 | |||
| AK148350.1 | - | 4 | 1262 | |||
| AK160906.1 | - | 4 | 1232 | |||
| BC006711.1 | - | 4 | 1155 | 3584570 | ||

Slc25a29
Genomic Map