siRNA Details
| Gene Symbol: Ube2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145420.2 | NP_663395.1 | 4 | 285 | ||
| GenBank mRNA | AK043167.1 | - | 3 | 2504 | ||
| AK144468.1 | - | 4 | 240 | |||
| AK148364.1 | - | 4 | 273 | |||
| BC019464.1 | - | 4 | 256 | 4205941 | ||
| BY111155.1 | - | 4 | 287 | |||

Ube2d1
Genomic Map