siRNA Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291282.1 | NP_001278211.1 | 4, 5 | 347 | ||
| NM_145375.4 | NP_663350.2 | 4, 5 | 855 | |||
| GenBank mRNA | AK046714.1 | - | 4, 5 | 756 | ||
| AK083132.1 | - | 4, 5 | 858 | |||
| AK135100.1 | - | 4, 5 | 837 | |||
| AK136581.1 | - | 4, 5 | 348 | |||
| AK163621.1 | - | 4, 5 | 780 | |||
| AK172109.1 | - | 4, 5 | 771 | |||
| AK172145.1 | - | 4, 5 | 756 | |||
| BC023123.1 | - | 4, 5 | 737 | 3995280 | ||

Tm6sf1
Genomic Map