siRNA Details
| Gene Symbol: Fam193b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145382.4 | NP_663357.2 | 8 | 2814 | ||
| GenBank mRNA | AK040568.1 | - | 2 | 942 | ||
| AK155237.1 | - | 8 | 2808 | |||
| AK173303.1 | - | 8 | 1663 | |||
| BC021381.1 | - | 2 | 404 | 5043282 | ||
| BC023015.1 | - | 2 | 346 | 5364758 | ||
| BC025483.1 | - | 8 | 1673 | 5150988 | ||
| BC051255.1 | - | 2 | 3262 | |||

Fam193b
Genomic Map