siRNA Details
| Gene Symbol: Slc44a3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145394.3 | NP_663369.2 | 4 | 552 | ||
| GenBank mRNA | AK145169.1 | - | 2 | 164 | ||
| BC010552.1 | - | 3 | 320 | 3582543 | ||
| BC025548.1 | - | 4 | 550 | 5320841 | ||
| BY735109.1 | - | 2 | 164 | |||
| CJ130557.1 | - | 1 | 53 | |||

Slc44a3
Genomic Map