siRNA Details
| Gene Symbol: IL12A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000882.3 | NP_000873.2 | 2 | 428 | ||
| GenBank mRNA | AF101062.1 | - | 2 | 207 | ||
| AF180562.1 | - | 2 | 266 | |||
| BC104982.1 | - | 2 | 360 | 8143985 | ||
| BC104984.1 | - | 2 | 360 | 8143987 | ||
| BG702253.1 | - | 2 | 521 | |||
| M65271.1 | - | 2 | 274 | |||
| M65291.1 | - | 2 | 308 | |||

IL12A
Genomic Map