siRNA Details
| Gene Symbol: Mfap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145426.2 | NP_663401.2 | 2, 3 | 523 | ||
| NM_180599.1 | NP_850930.1 | 2, 3 | 487 | |||
| GenBank mRNA | AK042988.1 | - | 2, 3 | 471 | ||
| AK076071.1 | - | 2, 3 | 489 | |||
| AK155122.1 | - | 2, 3 | 501 | |||
| BC006828.1 | - | 2, 3 | 482 | 3597878 | ||
| CF174749.1 | - | 2, 3 | 487 | |||

Mfap3
Genomic Map