siRNA Details
| Gene Symbol: Wbscr17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145218.3 | NP_660253.2 | 6 | 1931 | ||
| GenBank mRNA | AB175684.1 | - | 6 | 1009 | ||
| AF467979.1 | - | 6 | 1212 | |||
| AK035817.1 | - | 6 | 1685 | |||
| AK048758.1 | - | 6 | 1676 | |||
| AK051281.1 | - | 6 | 2331 | |||
| AK087726.1 | - | 2501 | ||||
| BC052469.1 | - | 6 | 1327 | 6810287 | ||
| BC069177.1 | - | 6 | 1931 | |||
| BC158110.1 | - | 6 | 1958 | 9088184 | ||

Wbscr17
Genomic Map