siRNA Details
| Gene Symbol: Tm6sf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001291282.1 | NP_001278211.1 | 10 | 1184 | ||
| NM_145375.4 | NP_663350.2 | 10 | 1692 | |||
| GenBank mRNA | AK083132.1 | - | 10 | 1695 | ||
| AK162572.1 | - | 8 | 1356 | |||
| AK163621.1 | - | 10 | 1617 | |||
| AK172109.1 | - | 10 | 1608 | |||
| BC023123.1 | - | 10 | 1574 | 3995280 | ||

Tm6sf1
Genomic Map