siRNA Details
| Gene Symbol: Best2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130194.1 | NP_001123666.1 | 7, 8 | 960 | ||
| GenBank mRNA | AY450428.1 | - | 7, 8 | 1202 | ||
| BC019528.1 | - | 6, 7 | 934 | 4208860 | ||
| BC031186.1 | - | 6, 7 | 921 | 4989959 | ||
| BC036157.1 | - | 7, 8 | 960 | 4982413 | ||
| BC036163.1 | - | 7, 8 | 1001 | 4984410 | ||

Best2
Genomic Map